Tisk…

Anti-Filamin A (phospho S2152) + Filamin B (phospho S2107) Rabbit Monoclonal Antibody [clone: EP2310AY]

Dodavatel: Abcam
AB75978-100 AB75978-1ML
ABCAAB75978-100EA 16750 CZK
ABCAAB75978-100 ABCAAB75978-1ML
Anti-Filamin A (phospho S2152) + Filamin B (phospho S2107) Rabbit Monoclonal Antibody [clone: EP2310AY]
Protilátky
Rabbit monoclonal [EP2310AY] to Filamin A (phospho S2152) + Filamin B (phospho S2107).

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Filamin A (phospho S2152) + Filamin B (phospho S2107)
Clonality: Monoclonal
Clone: EP2310AY
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR