Tisk…
Anti-CD3 zeta (phospho Y142) Rabbit Monoclonal Antibody [clone: EP265(2)Y]
Dodavatel: Abcam
AB68235-100 AB68235-20UL AB68235-1ML
ABCAAB68235-100EA 15860 CZK
ABCAAB68235-100 ABCAAB68235-20UL ABCAAB68235-1ML
Anti-CD3 zeta (phospho Y142) Rabbit Monoclonal Antibody [clone: EP265(2)Y]
Protilátky
Rabbit monoclonal [EP265(2)Y] to CD3 zeta (phospho Y142).
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD3 zeta
Clonality: Monoclonal
Clone: EP265(2)Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human