Tisk…

Anti-PAK1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB99448-100 ORB99448-20
BIRBORB99448-100EA 0 CZK
BIRBORB99448-100 BIRBORB99448-20
Anti-PAK1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: PAK1 (p21 protein (Cdc42/Rac)-activated kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR