Tisk…

Anti-CHEK1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB99317-100 ORB99317-20
BIRBORB99317-100EA 0 CZK
BIRBORB99317-100 BIRBORB99317-20
Anti-CHEK1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000

Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR