Tisk…

Anti-HIF1A Mouse Monoclonal Antibody

Dodavatel: Biorbyt
ORB67372-100 ORB67372-25
BIRBORB67372-100EA 0 CZK
BIRBORB67372-100 BIRBORB67372-25
Anti-HIF1A Mouse Monoclonal Antibody
Protilátky
Immunochemistry: 1:1000-1:8000

Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR