Tisk…

Anti-USO1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB136561-100 ORB136561-200 ORB136561-50
BIRBORB136561-100EA 0 CZK
BIRBORB136561-100 BIRBORB136561-200 BIRBORB136561-50
Anti-USO1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: USO1 (USO1 vesicle transport factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR