Tisk…

Anti-SLC5A1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB136312-100 ORB136312-200 ORB136312-50
BIRBORB136312-100EA 0 CZK
BIRBORB136312-100 BIRBORB136312-200 BIRBORB136312-50
Anti-SLC5A1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SLC5A1 (solute carrier family 5 (sodium/glucose cotransporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR