Tisk…

Anti-CCL17 Rabbit polyclonal antibody

Dodavatel: Biorbyt
ORB136281-100 ORB136281-200 ORB136281-50
BIRBORB136281-100EA 0 CZK
BIRBORB136281-100 BIRBORB136281-200 BIRBORB136281-50
Anti-CCL17 Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: CCL17 (chemokine (C-C motif) ligand 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR