Tisk…

Anti-NOTCH3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB135997-200 ORB135997-100 ORB135997-50
BIRBORB135997-200EA 0 CZK
BIRBORB135997-200 BIRBORB135997-100 BIRBORB135997-50
Anti-NOTCH3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NOTCH3 (notch 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR