Tisk…

Anti-SLC25A4 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB135052-50 ORB135052-100 ORB135052-200
BIRBORB135052-50EA 0 CZK
BIRBORB135052-50 BIRBORB135052-100 BIRBORB135052-200
Anti-SLC25A4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SLC25A4 (solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR