Tisk…

Anti-STAC2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB128785-100 ORB128785-200
BIRBORB128785-100EA 0 CZK
BIRBORB128785-100 BIRBORB128785-200
Anti-STAC2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: STAC2 (SH3 and cysteine rich domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR