Tisk…

Anti-RPS20 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB128674-100 ORB128674-200
BIRBORB128674-100EA 0 CZK
BIRBORB128674-100 BIRBORB128674-200
Anti-RPS20 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RPS20 (ribosomal protein S20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR