Tisk…

Anti-MSH2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB128403-100 ORB128403-200
BIRBORB128403-100EA 0 CZK
BIRBORB128403-100 BIRBORB128403-200
Anti-MSH2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MSH2 (mutS homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR