Tisk…

Anti-MINPP1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB128364-100 ORB128364-200
BIRBORB128364-100EA 0 CZK
BIRBORB128364-100 BIRBORB128364-200
Anti-MINPP1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MINPP1 (multiple inositol-polyphosphate phosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR