Tisk…

Anti-GIT1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB128107-100 ORB128107-200
BIRBORB128107-100EA 0 CZK
BIRBORB128107-100 BIRBORB128107-200
Anti-GIT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GIT1 (G protein-coupled receptor kinase interacting ArfGAP 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR