Tisk…
Anti-GFRA1 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB128105-100 ORB128105-200
BIRBORB128105-100EA 0 CZK
BIRBORB128105-100 BIRBORB128105-200
Anti-GFRA1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GFRA1 (GDNF family receptor alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat