Tisk…

Anti-CD3 zeta Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB127821-100 ORB127821-200
BIRBORB127821-100EA 0 CZK
BIRBORB127821-100 BIRBORB127821-200
Anti-CD3 zeta Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CD3 zeta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR