Tisk…
Anti-PDCD4 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB127501-100 ORB127501-200
BIRBORB127501-100EA 0 CZK
BIRBORB127501-100 BIRBORB127501-200
Anti-PDCD4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PDCD4 (programmed cell death 4 (neoplastic transformation inhibitor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat