Tisk…

Anti-PAK1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB127461-100 ORB127461-200
BIRBORB127461-100EA 0 CZK
BIRBORB127461-100 BIRBORB127461-200
Anti-PAK1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PAK1 (p21 protein (Cdc42/Rac)-activated kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR