Tisk…

Anti-RAB31 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB127379-100 ORB127379-200
BIRBORB127379-100EA 0 CZK
BIRBORB127379-100 BIRBORB127379-200
Anti-RAB31 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RAB31 (RAB31, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR