Tisk…

Anti-USP13 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB127312-100 ORB127312-200
BIRBORB127312-100EA 0 CZK
BIRBORB127312-100 BIRBORB127312-200
Anti-USP13 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: USP13 (ubiquitin specific peptidase 13 (isopeptidase T-3))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR