Tisk…

Anti-TAF1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB127258-100 ORB127258-200
BIRBORB127258-100EA 0 CZK
BIRBORB127258-100 BIRBORB127258-200
Anti-TAF1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TAF1 (TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR