Tisk…

Anti-NOX1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB127111-100 ORB127111-200
BIRBORB127111-100EA 0 CZK
BIRBORB127111-100 BIRBORB127111-200
Anti-NOX1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NOX1 (NADPH oxidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR