Tisk…

Anti-DDX19B Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB126863-100 ORB126863-200
BIRBORB126863-100EA 0 CZK
BIRBORB126863-100 BIRBORB126863-200
Anti-DDX19B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DDX19B (DEAD (Asp-Glu-Ala-Asp) box polypeptide 19B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR