Tisk…

Anti-RPS6KB2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB126703-100 ORB126703-200
BIRBORB126703-100EA 0 CZK
BIRBORB126703-100 BIRBORB126703-200
Anti-RPS6KB2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR