Tisk…

Anti-ZAK Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB126661-100 ORB126661-200
BIRBORB126661-100EA 0 CZK
BIRBORB126661-100 BIRBORB126661-200
Anti-ZAK Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ZAK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR