Tisk…

Anti-COL3A1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB126562-100 ORB126562-200
BIRBORB126562-100EA 0 CZK
BIRBORB126562-100 BIRBORB126562-200
Anti-COL3A1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: COL3A1 (collagen, type III, alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR