Tisk…

Anti-MMP3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB126558-100 ORB126558-200
BIRBORB126558-100EA 0 CZK
BIRBORB126558-100 BIRBORB126558-200
Anti-MMP3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MMP3 (matrix metallopeptidase 3 (stromelysin 1, progelatinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR