Tisk…

Anti-TUT1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB126464-100 ORB126464-200
BIRBORB126464-100EA 0 CZK
BIRBORB126464-100 BIRBORB126464-200
Anti-TUT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TUT1 (terminal uridylyl transferase 1, U6 snRNA-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR