Tisk…

Anti-SLC30A1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB126369-100 ORB126369-200
BIRBORB126369-100EA 0 CZK
BIRBORB126369-100 BIRBORB126369-200
Anti-SLC30A1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SLC30A1 (solute carrier family 30 (zinc transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR