Tisk…

Anti-MOS Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB125971-100 ORB125971-200
BIRBORB125971-100EA 0 CZK
BIRBORB125971-100 BIRBORB125971-200
Anti-MOS Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MOS (v-mos Moloney murine sarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR