Tisk…

Anti-MDC1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB125952-100 ORB125952-200
BIRBORB125952-100EA 0 CZK
BIRBORB125952-100 BIRBORB125952-200
Anti-MDC1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MDC1 (mediator of DNA-damage checkpoint 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR