Tisk…

Anti-MB Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB125950-100 ORB125950-200
BIRBORB125950-100EA 0 CZK
BIRBORB125950-100 BIRBORB125950-200
Anti-MB Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MB (myoglobin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR