Tisk…

Anti-EPN3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB125720-100 ORB125720-200
BIRBORB125720-100EA 0 CZK
BIRBORB125720-100 BIRBORB125720-200
Anti-EPN3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EPN3 (epsin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR