Tisk…
Anti-CD300LF Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB125532-100 ORB125532-200
BIRBORB125532-100EA 0 CZK
BIRBORB125532-100 BIRBORB125532-200
Anti-CD300LF Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CD300LF (CD300 molecule-like family member f)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human