Tisk…

Anti-ADCK2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB125389-100 ORB125389-200
BIRBORB125389-100EA 0 CZK
BIRBORB125389-100 BIRBORB125389-200
Anti-ADCK2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ADCK2 (aarF domain containing kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR