Tisk…

Anti-LAT Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB125343-100 ORB125343-200
BIRBORB125343-100EA 0 CZK
BIRBORB125343-100 BIRBORB125343-200
Anti-LAT Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR