Tisk…

Anti-ABCB7 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB125324-100 ORB125324-200
BIRBORB125324-100EA 0 CZK
BIRBORB125324-100 BIRBORB125324-200
Anti-ABCB7 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ABCB7 (ATP-binding cassette, sub-family B (MDR/TAP), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR