Tisk…

Anti-NF2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB106418-100 ORB106418-20
BIRBORB106418-100EA 0 CZK
BIRBORB106418-100 BIRBORB106418-20
Anti-NF2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:2000

Type: Primary
Antigen: NF2 (neurofibromin 2 (merlin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR