Tisk…

Anti-JAK2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB106305-100 ORB106305-20
BIRBORB106305-100EA 0 CZK
BIRBORB106305-100 BIRBORB106305-20
Anti-JAK2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: JAK2 (Janus kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR