Tisk…
Anti-USO1 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB101624-100 ORB101624-200
BIRBORB101624-100EA 0 CZK
BIRBORB101624-100 BIRBORB101624-200
Anti-USO1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: USO1 (USO1 vesicle transport factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat