Tisk…
Anti-MOS Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB101270-100 ORB101270-20 ORB101270-200 ORB101270-5
BIRBORB101270-100EA 0 CZK
BIRBORB101270-100 BIRBORB101270-20 BIRBORB101270-200 BIRBORB101270-5
Anti-MOS Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: MOS (v-mos Moloney murine sarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat