Tisk…

Anti-BOK Rabbit polyclonal antibody

Dodavatel: Biorbyt
ORB101174-100 ORB101174-200
BIRBORB101174-100EA 0 CZK
BIRBORB101174-100 BIRBORB101174-200
Anti-BOK Rabbit polyclonal antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: BOK (BCL2-related ovarian killer)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR