Tisk…

Anti-AK2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB100984-100 ORB100984-200
BIRBORB100984-100EA 0 CZK
BIRBORB100984-100 BIRBORB100984-200
Anti-AK2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: AK2 (adenylate kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR