Tisk…
Anti-CNNM2 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
ORB100454-100 ORB100454-200
BIRBORB100454-100EA 0 CZK
BIRBORB100454-100 BIRBORB100454-200
Anti-CNNM2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: CNNM2 (cyclin M2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat