Tisk…

Anti-SLC52A1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB100328-100 ORB100328-200
BIRBORB100328-100EA 0 CZK
BIRBORB100328-100 BIRBORB100328-200
Anti-SLC52A1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLC52A1 (solute carrier family 52 (riboflavin transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR