Tisk…

Anti-SDC1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB49138-100 ORB49138-200
BIRBORB49138-100EA 0 CZK
BIRBORB49138-100 BIRBORB49138-200
Anti-SDC1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SDC1 (syndecan 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR