Tisk…

Anti-UBE2S Mouse Monoclonal Antibody

Dodavatel: Biorbyt
ORB22796-100 ORB22796-20 ORB22796-5
BIRBORB22796-100EA 0 CZK
BIRBORB22796-100 BIRBORB22796-20 BIRBORB22796-5
Anti-UBE2S Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: UBE2S (ubiquitin-conjugating enzyme E2S)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR