Tisk…

Anti-UBE2C Goat Polyclonal Antibody

Dodavatel: Biorbyt
ORB18433-100 ORB18433-20
BIRBORB18433-100EA 0 CZK
BIRBORB18433-100 BIRBORB18433-20
Anti-UBE2C Goat Polyclonal Antibody
Protilátky
ELISA: 1:32000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: UBE2C (ubiquitin-conjugating enzyme E2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR