Tisk…

Anti-PEG3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
ORB13644-100 ORB13644-200
BIRBORB13644-100EA 0 CZK
BIRBORB13644-100 BIRBORB13644-200
Anti-PEG3 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: PEG3 (paternally expressed 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR